Learn More
EpCAM/TROP1 Rat anti-Mouse, Alexa Fluor 594, Clone: G8.8R, R&D Systems™
Rat MonoclonalAntibody
Manufacturer:  R&D Systems FAB8998T100UG
Catalog No. FAB8998T100
Add to cart
This item is not returnable. View return policy
Description
Description
Recombinant Monoclonal Antibody.
Specifications
Specifications
EpCAM/TROP1
Monoclonal
Alexa Fluor 594
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Rat
IgG2a
RUO
Primary
4072
Mouse
Flow Cytometry
G8.8R
Flow Cytometry 0.25-1 ug/10^6 cells
17-1A, 323/A3, ACSTD1, antigen identified by monoclonal AUA1, CD326, CD326 antigen, Cell surface glycoprotein Trop-1, chromosome 4, surface marker (35kD glycoprotein), DIAR5, EGP, EGP-2, EGP314, EGP40, EpCAM, epithelial cell adhesion molecule, Epithelial cell surface antigen, Epithelial glycoprotein, Epithelial glycoprotein 314, ESA, GA733-2, GA733-2EGP34, gp40, hEGP314, HNPCC8, KS 1/4 antigen, KS1/4, KSAHEA125, M1S2, M4S1, M4S1Ly74, Major gastrointestinal tumor-associated protein GA733-2, MIC18MH99, MOC31, TACST-1, TACSTD1, TROP1, TROP1CD326, Tumor-associated calcium signal transducer 1CO-17A
TE-71 Thymic epithelial cell line
100 ug
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Monoclonal
Detects mouse EpCAM/TROP-1.
SDS
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only (RUO)