Learn More
EpCAM/TROP1 Rat anti-Mouse, Alexa Fluor 647, Clone: G8.8R, R&D Systems™
Rat Monoclonal Antibody
Manufacturer:  R&D Systems FAB8998R025
Catalog No. FAB8998R025
Add to cart
This item is not returnable. View return policy
Description
Description
EpCAM/TROP1 Monoclonal antibody FAB8998R025 Specifically detects EpCAM/TROP1 in Mouse samples. It is validated for Flow Cytometry
Specifications
Specifications
EpCAM/TROP1
G8.8R
Flow Cytometry 5 uL/10^6 cells
Purified
Epcam
TE-71 Thymic epithelial cell line
Protein A or G purified from cell culture supernatant
RUO
Primary
4072
Mouse
Flow Cytometry
Alexa Fluor 647
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
17-1A, 323/A3, ACSTD1, antigen identified by monoclonal AUA1, CD326, CD326 antigen, Cell surface glycoprotein Trop-1, chromosome 4, surface marker (35kD glycoprotein), DIAR5, EGP, EGP-2, EGP314, EGP40, EpCAM, epithelial cell adhesion molecule, Epithelial cell surface antigen, Epithelial glycoprotein, Epithelial glycoprotein 314, ESA, GA733-2, GA733-2EGP34, gp40, hEGP314, HNPCC8, KS 1/4 antigen, KS1/4, KSAHEA125, M1S2, M4S1, M4S1Ly74, Major gastrointestinal tumor-associated protein GA733-2, MIC18MH99, MOC31, TACST-1, TACSTD1, TROP1, TROP1CD326, Tumor-associated calcium signal transducer 1CO-17A
Rat
IgG2a
25 Tests
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Monoclonal
Detects mouse EpCAM/TROP-1 in flow cytometry.