Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EPCR Mouse anti-Human, Alexa Fluor™ 647, Clone: OTI12H5, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP271607AF647

Catalog No. N271607A647


Add to cart

Description

Description

EPCR Monoclonal antibody specifically detects EPCR in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications

Specifications

EPCR
OTI12H5
Western Blot, Immunohistochemistry
Affinity Purified, Conjugated
PROCR
Recombinant protein fragment corresponding to amino acids 1-210 of human EPCR (NP_006395) produced in HEK293T cells.
Immunogen affinity purified
Store at 4°C in the dark.
Monoclonal
Human
Immunohistochemistry, Western Blot
Alexa Fluor 647
50mM Sodium Borate with 0.05% Sodium Azide
APC receptor, CCCA, CCD41centrosome-associated protein, CD201 antigen, centrocyclin, Endothelial cell protein C receptor, endothelial protein C receptor, EPCRMGC23024, protein C receptor, endothelial
Mouse
IgG2b
0.1 mL
Primary
10544.0
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit