Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EPGN Rat anti-Mouse, Alexa Fluor™ 532, Clone: RM0228-2L47, Novus Biologicals™

Rat Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP212284AF532

Catalog No. NP212284A32


Add to cart

Description

Description

EPGN Monoclonal antibody specifically detects EPGN in Mouse samples. It is validated for Western Blot.
Specifications

Specifications

EPGN
RM0228-2L47
Western Blot
epigen, epithelial mitogen homolog (mouse), PRO9904, UNQ3072/PRO9904
Rat
IgG2
Store at 4°C in the dark.
Monoclonal
Mouse
Western Blot
Alexa Fluor™ 532
50 mM sodium borate with 0.05% sodium azide
EPGN
Mouse EPGN
0.1 mL
Primary
255324
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit