Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EphA2 Mouse anti-Human, Janelia Fluor 549, Clone: 3D7, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP123018JF549

Catalog No. NBP12318549

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

EphA2Monoclonalantibody specifically detects EphA2 in Human samples.
Specifications

Specifications

EphA2
3D7
Immunohistochemistry, Immunohistochemistry-Paraffin
Purified
EPHA2
Recombinant human EPH Receptor A2 (559-976aa) purified from E. coli
Protein G purified
Cancer, Protein Kinase
Primary
1969
Human
Immunohistochemistry, Immunohistochemistry (Paraffin)
Janelia Fluor 549
50mM Sodium Borate with 0.05% Sodium Azide
ARCC2, EC 2.7.10, EC 2.7.10.1, ECKepithelial cell receptor protein tyrosine kinase, EPH receptor A2, EphA2, ephrin type-A receptor 2, Epithelial cell kinase, soluble EPHA2 variant 1, Tyrosine-protein kinase receptor ECK
Mouse
IgG2
0.1mL
Store at 4°C in the dark.
Monoclonal
Anti-human EPH Receptor A2 mAb, clone 3D7, is derived from hybridization of mouse F0 myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human EPH Receptor A2 protein.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit