Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EphA2 Rabbit anti-Human, Alexa Fluor™ 750, Clone: 1C1, Novus Biologicals™

Rabbit Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP252677AF750

Catalog No. NP22677A70


Add to cart

Description

Description

EphA2 Monoclonal antibody specifically detects EphA2 in Human samples. It is validated for Western Blot,Flow Cytometry,ELISA,Immunofluorescence.
Specifications

Specifications

EphA2
1C1
Western Blot, Flow Cytometry, ELISA, Immunofluorescence
ARCC2, EC 2.7.10, EC 2.7.10.1, ECKepithelial cell receptor protein tyrosine kinase, EPH receptor A2, EphA2, ephrin type-A receptor 2, Epithelial cell kinase, soluble EPHA2 variant 1, Tyrosine-protein kinase receptor ECK
Rabbit
IgG Kappa
Store at 4°C in the dark.
Monoclonal
1C1 binds specifically to the amino-termianal LBD (ligand-binding domain) region of the EphA2 ectodomain (IgG1 KD ~0.8 nM; Fab KD ~140 nM) and not to other related human Eph receptors such as EphA1, EphA2, and EphA4, or any EphB receptor family members.
ELISA, Flow Cytometry, Immunofluorescence, Western Blot
Alexa Fluor™ 750
50 mM sodium borate with 0.05% sodium azide
EPHA2
1C1 was isolated after screening of a non-immunized human Fab phage library. Selection was based on the ability to bind soluble human recombinant EphA2 (residues 183-510). The Fab was subsequently converted to full length human IgG1.
0.1 mL
Primary
1969
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit