Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EphA2 Rat anti-Mouse, Alexa Fluor 350, Clone: 233720, R&D Systems™
Rat Monoclonal Antibody
Supplier: R&D Systems FAB639U100UG
This item is not returnable.
View return policy
Description
EphA2 Monoclonal specifically detects EphA2 in Mouse samples. It is validated for Flow Cytometry.Specifications
EphA2 | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
AAA82113 | |
EPHA2 | |
Mouse myeloma cell line NS0-derived recombinant mouse EphA2 Ala22-Ala535 (predicted) Accession # AAA82113 | |
RUO | |
1969 | |
Mouse | |
IgG2b |
Flow Cytometry | |
233720 | |
Alexa Fluor 350 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
ARCC2, EC 2.7.10, EC 2.7.10.1, Eck, ECKepithelial cell receptor protein tyrosine kinase, EPH receptor A2, EphA2, ephrin type-A receptor 2, Epithelial cell kinase, Myk2, Sek2, soluble EPHA2 variant 1, Tyrosine-protein kinase receptor ECK | |
Rat | |
100 μg | |
Primary | |
Detects mouse EphA2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human EphA1, recombinant mouse (rm) EphA3, 4, 6, 7, 8, recombinant rat EphA5, rmEphB1, 2, 3, 4, or 6 is observed. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)