Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EphA2 Rat anti-Mouse, Alexa Fluor 647, Clone: 8F21, Novus Biologicals™

Rat Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NB11093602AF647

Catalog No. NBT93602647

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

EphA2
8F21
Western Blot
Purified
EPHA2
Recombinant mouse EphA2 extracellular domain
Protein G purified
Cancer, Protein Kinase
Primary
1969
Mouse
Western Blot
Alexa Fluor 647
50mM Sodium Borate with 0.05% Sodium Azide
ARCC2, EC 2.7.10, EC 2.7.10.1, ECKepithelial cell receptor protein tyrosine kinase, EPH receptor A2, EphA2, ephrin type-A receptor 2, Epithelial cell kinase, soluble EPHA2 variant 1, Tyrosine-protein kinase receptor ECK
Rat
IgG1
0.1 ml
Store at 4C in the dark.
Monoclonal
This was selected for its ability to detect mouse EphA2 in western blots. In this format, this does not cross-react with recombinant mouse EphA3, 4, 6, 7, 8, and EphB1, 2, 3, 4, or 6.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit