Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EphA3 Rat anti-Mouse, Alexa Fluor 488, Clone: 7C13, Novus Biologicals™

Rat Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NB11093603AF488

Catalog No. NBT93603488


Add to cart

Description

Description

EphA3 Monoclonal antibody specifically detects EphA3 in Mouse samples. It is validated for Western Blot.
Specifications

Specifications

EphA3
7C13
Western Blot
Purified
EPHA3
Recombinant mouse EphA3 extracellular domain
Protein G purified
Protein Kinase
Primary
2042
Mouse
Western Blot
Alexa Fluor 488
50mM Sodium Borate with 0.05% Sodium Azide
EC 2.7.10, EC 2.7.10.1, EK4, EPH receptor A3, EphA3, EPH-like kinase 4, ephrin type-A receptor 3, ETK, hEK4, HEKETK1eph-like tyrosine kinase 1, human embryo kinase 1, TYRO4, TYRO4 protein tyrosine kinase, Tyrosine-protein kinase receptor ETK1, Tyrosine-protein kinase TYRO4
Rat
IgG2
0.1 ml
Store at 4C in the dark.
Monoclonal
This was selected for its ability to detect mouse EphA3 in western blots. In this format, this does not cross-react with recombinant mouse EphA2, EphA6, EphA4, EphA7, EphA8 and EphB2, EphB3 or EphB6.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit