Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EphA4 Rabbit anti-Mouse, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP19836320UL
Description
EphA4 Polyclonal specifically detects EphA4 in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
EphA4 | |
Polyclonal | |
Western Blot 1:1000, Immunohistochemistry 1:10-1:500 | |
NP_031962 | |
EPHA4 | |
The immunogen for this antibody is Eph receptor A4. Peptide sequence VISRRRSKYSKAKQEADEEKHLNQGVRTYVDPFTYEDPNQAVREFAKEID. | |
Affinity Purified | |
RUO | |
2043 | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot, Immunohistochemistry | |
Unconjugated | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
EC 2.7.10, EC 2.7.10.1, EK8, EPH receptor A4, EphA4, EPH-like kinase 8, ephrin type-A receptor 4, HEK8, receptor protein-tyrosine kinase HEK8, SEK, TYRO1, TYRO1 protein tyrosine kinase, Tyrosine-protein kinase receptor SEK, Tyrosine-protein kinase TYRO1 | |
Rabbit | |
110 kDa | |
20 μL | |
Primary | |
Mouse | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title