Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EphA4 Rat anti-Mouse, Alexa Fluor 647, Clone: 11C13, Novus Biologicals™

Rat Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NB11093604AF647

Catalog No. NBT93604647


Add to cart

Description

Description

EphA4 Monoclonal antibody specifically detects EphA4 in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

EphA4
11C13
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Purified
EPHA4
Recombinant mouse EPH Receptor A4 (11C13) extracellular domain
Protein G purified
Protein Kinase
Primary
2043
Mouse
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Alexa Fluor 647
50mM Sodium Borate with 0.05% Sodium Azide
EC 2.7.10, EC 2.7.10.1, EK8, EPH receptor A4, EphA4, EPH-like kinase 8, ephrin type-A receptor 4, HEK8, receptor protein-tyrosine kinase HEK8, SEK, TYRO1, TYRO1 protein tyrosine kinase, Tyrosine-protein kinase receptor SEK, Tyrosine-protein kinase TYRO1
Rat
IgG1
0.1 ml
Store at 4C in the dark.
Monoclonal
This was selected for its ability to detect mouse EphA4 in western blots. In this format, this does not cross-react with recombinant mouse EphA2, EphA6, EphA7, EphA8 and EphB2, EphB3, EphB4 or EphB6.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit