Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EphA5 Polyclonal Antibody
EphA5 Polyclonal Antibody
Supplier: Thermo Scientific PA595282
Description
EphA5 Polyclonal Antibody for Western Blot

Specifications
EphA5 | |
Polyclonal | |
Unconjugated | |
EPHA5 | |
AI854630; ARCC2; AW125296; brain-specific kinase; BSK; CEK7; CEK-7; CTPA; CTPP1; ECK; eck-like sequence 1; EHK1; EHK-1; EK7; Ekh1; Els1; Els1.; epa5; EPH homology kinase 1; Eph receptor A5; EPHA5; EPH-like kinase 7; Ephrin A2; Ephrin A5; Ephrin type-A receptor 5; EphrinA2; EphrinA5; HEK7; receptor protein-tyrosine kinase HEK7; Rek7; TYRO4; tyrosine-protein kinase receptor EHK-1 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
13839, 2044, 79208 | |
-20°C | |
Lyophilized |
Western Blot | |
500 μg/mL | |
PBS with 4mg trehalose and 0.05mg sodium azide | |
P54756, P54757, Q60629 | |
EPHA5 | |
A synthetic peptide corresponding to a sequence at the C-terminus of human Eph receptor A5 (1001-1032aa TLEDLRRLGVTLVGHQKKIMNSLQEMKVQLVN). | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction