Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EphA6 Rat anti-Mouse, Alexa Fluor 700, Clone: 9J12, Novus Biologicals™

Rat Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NB11093605AF700

Catalog No. NBT93605700


Add to cart

Description

Description

EphA6 Monoclonal antibody specifically detects EphA6 in Mouse samples. It is validated for Western Blot.
Specifications

Specifications

EphA6
9J12
Western Blot
Purified
EPHA6
Recombinant mouse EPH Receptor A6 Antibody (9J12) extracellular domain
Protein G purified
Neuronal Cell Markers, Neuroscience, Neurotransmission
Primary
285220
Mouse
Western Blot
Alexa Fluor 700
50mM Sodium Borate with 0.05% Sodium Azide
DKFZp434C1418, EC 2.7.10, EC 2.7.10.1, EHK2, EHK-2, EPA6, EPH homology kinase 2, EPH receptor A6, ephrin receptor EphA6, ephrin type-A receptor 6, FLJ35246, PRO57066
Rat
IgG1
0.1 ml
Store at 4C in the dark.
Monoclonal
This was selected for its ability to detect mouse EPH Receptor A6 Antibody (9J12) in western blots. In this format, this does not cross-react with recombinant mouse EphA2, EphA3, EphA4, EphA7, EphA8 and EphB2, EphB3, EphB4 or EphB6.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit