Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EphA7 Rat anti-Mouse, Alexa Fluor 488, Clone: 60, Novus Biologicals™
Rat Monoclonal Antibody
Supplier: Novus Biologicals NB11093606AF488
Description
EphA7 Monoclonal specifically detects EphA7 in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
EphA7 | |
Monoclonal | |
Alexa Fluor 488 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
EPHA7 | |
Mouse EphA7 (extracellular domain) | |
0.1 mL | |
Primary | |
This was selected for its ability to detect mouse EphA7 in western blots. In this format, this does not cross-react with recombinant mouse EphA2, EphA3, EphA4, EphA6, EphA8 and EphB2, EphB3 or EphB4. | |
Store at 4C in the dark. | |
IgG1 |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
60 | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
EC 2.7.10, EC 2.7.10.1, EHK3, EHK-3, EK11, EPH homology kinase 3, Eph homology kinase-3, EPH receptor A7, EphA7, EPH-like kinase 11, ephrin type-A receptor 7, HEK11, receptor protein-tyrosine kinase HEK11, tyrosine-protein kinase receptor EHK-3 | |
Rat | |
Protein A or G purified | |
Neuronal Cell Markers, Neuroscience, Neurotransmission, Protein Kinase | |
2045 | |
Mouse | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title