Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EphA7 Rat anti-Mouse, DyLight 350, Clone: 60, Novus Biologicals™

Rat Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NB11093606UV

Catalog No. NBT93606UV


Add to cart

Description

Description

EphA7 Monoclonal antibody specifically detects EphA7 in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

EphA7
60
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Purified
EPHA7
Recombinant mouse EphA7 extracellular domain
Protein G purified
Neuronal Cell Markers, Neuroscience, Neurotransmission, Protein Kinase
Primary
2045
Mouse
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
DyLight 350
50mM Sodium Borate with 0.05% Sodium Azide
EC 2.7.10, EC 2.7.10.1, EHK3, EHK-3, EK11, EPH homology kinase 3, Eph homology kinase-3, EPH receptor A7, EphA7, EPH-like kinase 11, ephrin type-A receptor 7, HEK11, receptor protein-tyrosine kinase HEK11, tyrosine-protein kinase receptor EHK-3
Rat
IgG1
0.1 ml
Store at 4C in the dark.
Monoclonal
This was selected for its ability to detect mouse EphA7 in western blots. In this format, this does not cross-react with recombinant mouse EphA2, EphA3, EphA4, EphA6, EphA8 and EphB2, EphB3 or EphB4.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit