Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EphB4 Rabbit anti-Human, Clone: 1, Novus Biologicals™
Rabbit Monoclonal Antibody
$350.00 - $542.50
Specifications
Antigen | EphB4 |
---|---|
Clone | 1 |
Dilution | Flow Cytometry 1:25-1:100, ELISA 1:5000-1:10000 |
Applications | Flow Cytometry, ELISA |
Classification | Monoclonal |
Description
EphB4 Monoclonal specifically detects EphB4 in Human samples. It is validated for Flow Cytometry, ELISA.Specifications
EphB4 | |
Flow Cytometry 1:25-1:100, ELISA 1:5000-1:10000 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
EC 2.7.10, EC 2.7.10.1, EPH receptor B4, EphB4, ephrin type-B receptor 4, hepatoma transmembrane kinase, HTKephrin receptor EphB4, MYK1, soluble EPHB4 variant 1, soluble EPHB4 variant 2, soluble EPHB4 variant 3, Tyro11, Tyrosine-protein kinase receptor HTK, Tyrosine-protein kinase TYRO11 | |
Recombinant Human EphB4 protein | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
1 | |
Flow Cytometry, ELISA | |
Unconjugated | |
Rabbit | |
Protein Kinase | |
0.2 um filtered solution in PBS | |
2050 | |
IgG | |
Protein A purified |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title