Learn More
EPHB4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 208831AP
Catalog No. 50-173-3137
Add to cart
Description
Description
Ephrin receptors and their ligands, the ephrins, mediate numerous developmental processes, particularly in the nervous system. Based on their structures and sequence relationships, ephrins are divided into the ephrin-A (EFNA) class, which are anchored to the membrane by a glycosylphosphatidylinositol linkage, and the ephrin-B (EFNB) class, which are transmembrane proteins. The Eph family of receptors are divided into 2 groups based on the similarity of their extracellular domain sequences and their affinities for binding ephrin-A and ephrin-B ligands. Ephrin receptors make up the largest subgroup of the receptor tyrosine kinase (RTK) family. The protein encoded by this gene binds to ephrin-B2 and plays an essential role in vascular development.
Specifications
Specifications
EPHB4
0.21 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P54760, P54761
EPH receptor B4, EPHB4, Ephrin type B receptor 4, HTK, MYK1, TYRO11, Tyrosine protein kinase TYRO11
Rabbit
IgG
150 μL
-20°C
Primary
13846, 2050, 686310
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
EPHB4
Liquid
Ephb4
EPHB4 Fusion Protein Ag10033
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat