Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EphB4 Rat anti-Mouse, DyLight 680, Clone: 6A14, Novus Biologicals™

Rat Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NB11093607FR

Catalog No. NBT93607FR

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

EphB4
6A14
Western Blot
Purified
EPHB4
Recombinant mouse EPH Receptor B4 Antibody (6A14) extracellular domain
Protein G purified
Protein Kinase
Primary
2050
Mouse
Western Blot
DyLight 680
50mM Sodium Borate with 0.05% Sodium Azide
EC 2.7.10, EC 2.7.10.1, EPH receptor B4, EphB4, ephrin type-B receptor 4, hepatoma transmembrane kinase, HTKephrin receptor EphB4, MYK1, soluble EPHB4 variant 1, soluble EPHB4 variant 2, soluble EPHB4 variant 3, Tyro11, Tyrosine-protein kinase receptor HTK, Tyrosine-protein kinase TYRO11
Rat
IgG1
0.1 ml
Store at 4C in the dark.
Monoclonal
This was selected for its ability to detect mouse EPH Receptor B4 Antibody (6A14) in western blots. In this format, this does not cross-react with recombinant mouse EphA2, EphA3, EphA4, EphA6, EphA7 or EphB6.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit