Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ EphB6 Antibody Pair [HRP]
SDP

Rabbit Monoclonal Antibody Pair

$1219.00

Specifications

Assay Range 9.4 pg/mL
Assay Sensitivity 9.38-600 pg/mL
Assay Sandwich ELISA
Conjugate HRP
Product Type Antibody Pairs
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB010019
SDP
View Documents
Novus Biologicals™
NBP27949015PLATES
15 Plates
Each of 1 for $1,219.00
Add to Cart
 
Description

Description

This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human EphB6.
Specifications

Specifications

9.4 pg/mL
Sandwich ELISA
Antibody Pairs
Human
EC 2.7.10.1, EPH receptor B6, EphB6, ephrin type-B receptor 6, HEPTyrosine-protein kinase-defective receptor EPH-6, MGC129910, MGC129911
EPHB6
  • Standard: Each vial contains 8 ng of recombinant EphB6.
  • Capture Antibody: 1.0 mg/mL of rabbit anti-EphB6 monoclonal antibody (in PBS, pH 7.4).
  • Detection Antibody: 0.5 mg/mL rabbit anti-EphB6 monoclonal antibody conjugated to horseradish-peroxidase (HRP) (in PBS, 50 % glycerol, pH 7.4).
  • Protein Kinase
    EphB6
    9.38-600 pg/mL
    HRP
    This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human EphB6.
    Sandwich ELISA
    2051
    Rabbit
    RUO
    Storage is content dependent.
    1. Capture Antibody: Dilute to a working concentration of 2 μg/mL in CBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.5 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 0.6 ng/mL is recommended.
    SDS
    Documents

    Documents

    Product Certifications
    Provide Content Correction

    We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Cancel Submit