Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EphB6 Rat anti-Mouse, Alexa Fluor 405, Clone: 9B17, Novus Biologicals™

Rat Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NB11093608AF405

Catalog No. NBT93608405

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

EphB6
9B17
Western Blot
Purified
EPHB6
Recombinant mouse EPH Receptor B6 Antibody (9B17) extracellular domain
Protein G purified
Protein Kinase
Primary
2051
Mouse
Western Blot
Alexa Fluor 405
50mM Sodium Borate with 0.05% Sodium Azide
EC 2.7.10.1, EPH receptor B6, EphB6, ephrin type-B receptor 6, HEPTyrosine-protein kinase-defective receptor EPH-6, MGC129910, MGC129911
Rat
IgG2
0.1 ml
Store at 4C in the dark.
Monoclonal
This was selected for its ability to detect mouse EPH Receptor B6 Antibody (9B17) in western blots. In this format, this does not cross-react with recombinant mouse EphA2, EphA3, EphA4, EphA6, EphA7 EphA8, EphB2, EphB3 or EphB4.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit