Learn More
Ephrin-A1 Rat anti-Mouse, Alexa Fluor™ 532, Clone: RM0224-6D31, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP212277AF532
Catalog No. NP212277A32
Add to cart
Description
Description
Ephrin-A1 Monoclonal antibody specifically detects Ephrin-A1 in Mouse samples. It is validated for Western Blot.
Specifications
Specifications
Ephrin-A1
RM0224-6D31
Western Blot
ECKLG, EFL1, EPH-related receptor tyrosine kinase ligand 1, ephrin-A1, EPLG1TNF alpha-induced protein 4, Immediate early response protein B61, LERK1LERK-1, ligand of eph-related kinase 1, TNFAIP4B61, Tumor necrosis factor alpha-induced protein 4, tumor necrosis factor, alpha-induced protein 4
Rat
IgG2
Store at 4°C in the dark.
Monoclonal
Mouse
Western Blot
Alexa Fluor™ 532
50 mM sodium borate with 0.05% sodium azide
EFNA1
Mouse Ephrin A1
0.1 mL
Primary
1942