Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ephrin-A1 Rat anti-Mouse, Clone: RM0224-6D31, Novus Biologicals™

Rat Monoclonal Antibody

$177.16 - $493.00

Specifications

Antigen Ephrin-A1
Clone RM0224-6D31
Concentration LYOPH
Applications Western Blot
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP212277
View Documents
Novus Biologicals
NBP212277
0.1 mg
Each for $493.00
Add to cart
 
NB019140
View Documents
Novus Biologicals
NBP2122770.025MG
0.025 mg
Each for $177.16
Add to cart
 
Description

Description

Ephrin-A1 Monoclonal specifically detects Ephrin-A1 in Mouse samples. It is validated for Western Blot.
Specifications

Specifications

Ephrin-A1
LYOPH
Monoclonal
Purified
RUO
Mouse
ECKLG, EFL1, EPH-related receptor tyrosine kinase ligand 1, ephrin-A1, EPLG1TNF alpha-induced protein 4, Immediate early response protein B61, LERK1LERK-1, ligand of eph-related kinase 1, TNFAIP4B61, Tumor necrosis factor alpha-induced protein 4, tumor necrosis factor, alpha-induced protein 4
EFNA1
Primary
RM0224-6D31
Western Blot
Unconjugated
Rat
Cancer
P52793
1942
IgG2
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
SDS
Product Certifications

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit