Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ephrin-A2 Mouse anti-Human, Allophycocyanin/Cy7, Clone: OTI3E3, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270622APCCY7

Catalog No. N270622APC7


Add to cart

Description

Description

Ephrin-A2 Monoclonal antibody specifically detects Ephrin-A2 in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

Ephrin-A2
OTI3E3
Flow Cytometry
Affinity Purified, Conjugated
EFNA2
Full length recombinant protein of human Ephrin-A2 (NP_001396) produced in HEK293T cells.
Immunogen affinity purified
Store at 4°C in the dark. Do not freeze.
Monoclonal
Human
Flow Cytometry
APC-Cyanine7
PBS with 0.05% Sodium Azide
ELF-1, EPH-related receptor tyrosine kinase ligand 6, ephrin-A2, EPLG6HEK7 ligand, HEK7-L, HEK7-ligand, LERK6LERK-6, ligand of eph-related kinase 6
Mouse
IgG2b
0.1 mL
Primary
1943.0
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit