Learn More
Ephrin-A2 Rat anti-Mouse, Alexa Fluor™ 750, Clone: RM0225-7H45, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP212278AF750
Catalog No. NP212278A70
Add to cart
Description
Description
Ephrin-A2 Monoclonal antibody specifically detects Ephrin-A2 in Mouse samples. It is validated for Western Blot.
Specifications
Specifications
Ephrin-A2
RM0225-7H45
Western Blot
ELF-1, EPH-related receptor tyrosine kinase ligand 6, ephrin-A2, EPLG6HEK7 ligand, HEK7-L, HEK7-ligand, LERK6LERK-6, ligand of eph-related kinase 6
Rat
IgG2
Store at 4°C in the dark.
Monoclonal
Mouse
Western Blot
Alexa Fluor™ 750
50 mM sodium borate with 0.05% sodium azide
EFNA2
Mouse Ephrin A2
0.1 mL
Primary
1943