Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ephrin-A3 Mouse anti-Human, Alexa Fluor™ 405, Clone: OTI4A4, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270623AF405

Catalog No. N270623A405


Add to cart

Description

Description

Ephrin-A3 Monoclonal antibody specifically detects Ephrin-A3 in Human samples. It is validated for Immunohistochemistry.
Specifications

Specifications

Ephrin-A3
OTI4A4
Immunohistochemistry
Affinity Purified, Conjugated
EFNA3
Recombinant protein fragment corresponding to amino acids 23-214 of human Ephrin-A3 (NP_004943) produced in E.coli.
Immunogen affinity purified
Growth and Development, Neuronal Cell Markers
Primary
1944.0
Immunohistochemistry
Alexa Fluor 405
50mM Sodium Borate with 0.05% Sodium Azide
EFL2, EFL-2, EHK1-L, EPH-related receptor tyrosine kinase ligand 3, ephrin-A3, EPLG3EHK1 ligand, LERK3LERK-3, ligand of eph-related kinase 3
Mouse
IgG2b
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit