Learn More
Ephrin-A4 Mouse anti-Human, Alexa Fluor™ 750, Clone: MM0263-9H13, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP212280AF750
Catalog No. NP212280A70
Add to cart
Description
Description
Ephrin-A4 Monoclonal antibody specifically detects Ephrin-A4 in Human samples. It is validated for Western Blot,Immunohistochemistry,Immunohistochemistry (Paraffin).
Specifications
Specifications
Ephrin-A4
MM0263-9H13
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
EFL4, EPH-related receptor tyrosine kinase ligand 4, ephrin-A4, EPLG4MGC125826, LERK-4, LERK4FLJ57652, ligand of eph-related kinase 4
Mouse
IgG1
Store at 4°C in the dark.
Monoclonal
Human
Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Alexa Fluor™ 750
50 mM sodium borate with 0.05% sodium azide
EFNA4
Human Ephrin A4
0.1 mL
Primary
1945