Learn More
Ephrin A4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 196851AP
Catalog No. 50-173-2485
Add to cart
Description
Description
The antibody is specific to EFNA4.
Ephrin A4 is also known as LERK-4 or ELF-4. Ephrin-A4 binds EphA2, EphA3, EphA4, EphA5, EphA6, EphA7, and EphB1. To date, at least 14 members of the Eph receptor family and a family of 8 ligands have been identified. Ligands of Eph family receptors are structurally related membrane-bound proteins that can be subdivided into two major subclasses: ephrin-A and ephrin-B. Ligands in the ephrin-A subclass, including the prototype family member ephrin-A1 (B61), are membrane associated through glycosylphosphatidyl-inositollinkages; whereas ephrin-B subclass consists of ligands with transmembrane domains.
Specifications
Specifications
Ephrin A4
0.17 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O08542, P52798
EFL4, EFNA4, ephrin A4, EPLG4, LERK 4, LERK4
Rabbit
IgG
150 μL
-20°C
Primary
13639, 1945, 310643
Western Blot
Unconjugated
EFNA4
Liquid
Efna4
Peptide
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat