Learn More
Ephrin-A4 Rat anti-Mouse, Alexa Fluor™ 594, Clone: RM0226-8K23, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP212281AF594
Catalog No. NP212281A94
Add to cart
Description
Description
Ephrin-A4 Monoclonal antibody specifically detects Ephrin-A4 in Mouse samples. It is validated for Western Blot.
Specifications
Specifications
Ephrin-A4
RM0226-8K23
Western Blot
EFL4, EPH-related receptor tyrosine kinase ligand 4, ephrin-A4, EPLG4MGC125826, LERK-4, LERK4FLJ57652, ligand of eph-related kinase 4
Rat
IgG2
Store at 4°C in the dark.
Monoclonal
Mouse
Western Blot
Alexa Fluor™ 594
50 mM sodium borate with 0.05% sodium azide
EFNA4
Mouse Ephrin A4
0.1 mL
Primary
1945