Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ephrin-B1 Rat anti-Mouse, Alexa Fluor™ 750, Clone: RM0227-12A31, Novus Biologicals™

Rat Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP212282AF750

Catalog No. NP212282A70


Add to cart

Description

Description

Ephrin-B1 Monoclonal antibody specifically detects Ephrin-B1 in Mouse samples. It is validated for Western Blot.
Specifications

Specifications

Ephrin-B1
RM0227-12A31
Western Blot
EFL3, EFL-3, ELK ligand, Elk-L, ephrin-B1, LERK-2
Rat
IgG2
Store at 4°C in the dark.
Monoclonal
Mouse
Western Blot
Alexa Fluor™ 750
50 mM sodium borate with 0.05% sodium azide
EFNB1
Mouse Ephrin B
0.1 mL
Primary
1947
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit