Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ephrin-B2 Rabbit anti-Human, Clone: JM53-21, Novus Biologicals™

Rabbit Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP266821

Catalog No. NBP266821


Add to cart

Description

Description

Ephrin-B2 Monoclonal antibody specifically detects Ephrin-B2 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

Ephrin-B2
JM53-21
Unconjugated
Purified
ephrin B2, EPLG5, Htk-L, HTKL, LERK5
EFNB2
Recombinant Protein
Monoclonal
Protein A purified
Neuroscience, Signal Transduction
Human,Mouse,Rat
Immunocytochemistry,Immunofluorescence,Western Blot
1 mg/mL
Western Blot 1:500-1:1000, Immunocytochemistry/Immunofluorescence 1:50-1:200
TBS (pH7.4), 1%BSA, 40%Glycerol with 0.05% Sodium Azide
1948
Rabbit
IgG
Primary
100μL
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit