Learn More
Ephrin-B3 Mouse anti-Human, Alexa Fluor™ 594, Clone: MM0264- 12X1, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP212283AF594
Catalog No. NP212283A94
Add to cart
Description
Description
Ephrin-B3 Monoclonal antibody specifically detects Ephrin-B3 in Human samples. It is validated for Western Blot,Immunohistochemistry,Immunohistochemistry (Paraffin).
Specifications
Specifications
Ephrin-B3
MM0264- 12X1
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
EPH-related receptor tyrosine kinase ligand 8, Ephrin B3, ephrin-B3, EPLG8EPH-related receptor transmembrane ligand ELK-L3, LERK-8, LERK8EFL6
Mouse
IgG1
Store at 4°C in the dark.
Monoclonal
Human
Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Alexa Fluor™ 594
50 mM sodium borate with 0.05% sodium azide
EFNB3
Human Ephrin B3
0.1 mL
Primary
1949