Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EPHX1 Mouse anti-Human, Mouse, Rat, Monkey, DyLight 650, Clone: OTI3F10, Novus Biologicals
Mouse Monoclonal Antibody
Manufacturer: Novus Biologicals NBP270646C
Description
EPHX1 Monoclonal antibody specifically detects EPHX1 in Human, Mouse, Rat, Monkey samples. It is validated for Western Blot, Immunohistochemistry.Specifications
EPHX1 | |
OTI3F10 | |
Western Blot, Immunohistochemistry | |
Affinity Purified, Conjugated | |
EPHX1 | |
Recombinant protein fragment corresponding to amino acids 21-230 of human EPHX1 (NP_000111) produced in E.coli. | |
Immunogen affinity purified | |
Cancer | |
Primary | |
2052.0 |
Immunohistochemistry, Western Blot | |
DyLight 650 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
EC 3.3.2.9, EPHX, EPOX, Epoxide hydratase, epoxide hydrolase 1, epoxide hydrolase 1, microsomal (xenobiotic), MEH, Microsomal epoxide hydrolase | |
Mouse | |
IgG2b | |
0.1 mL | |
Store at 4°C in the dark. | |
Monoclonal | |
Human, Monkey, Mouse, Rat |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title