Learn More
EPHX2 Antibody (OTI1A6), DyLight 405, Novus Biologicals™
SDP
Mouse Monoclonal Antibody
Supplier:  Novus Biologicals NBP270648V
Catalog No. NBP270648V
Add to Cart
Description
Description
EPHX2 Monoclonal specifically detects EPHX2 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
EPHX2
Monoclonal
DyLight 405
50mM Sodium Borate with 0.05% Sodium Azide
EPHX2
Full length recombinant protein of human EPHX2 (NP_001970) produced in HEK293T cells.
0.1 mL
Primary
Human, Mouse
IgG1
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
OTI1A6
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
CEH, Cytosolic epoxide hydrolase, EC 3.3.2.10, Epoxide hydratase, epoxide hydrolase 2, epoxide hydrolase 2, cytoplasmic, epoxide hydrolase 2, cytosolic, epoxide hydrolase, soluble, SEH, Soluble epoxide hydrolase
Mouse
Affinity Purified
Lipid and Metabolism
2053.0
Store at 4°C in the dark.
Product Suggestions
Product Suggestions