Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EPHX2 Mouse anti-Human, Clone: OTI1A6, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA525222

Catalog No. PIMA525222


Add to cart

Description

Description

This gene encodes a member of the epoxide hydrolase family. The protein, found in both the cytosol and peroxisomes, binds to specific epoxides and converts them to the corresponding dihydrodiols. Mutations in this gene have been associated with familial hypercholesterolemia. Alternatively spliced transcript variants have been described.
Specifications

Specifications

EPHX2
Monoclonal
0.7 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
P34913
CEH;Cytosolic epoxide hydrolase;EPHX2;Epoxide hydratase;Epoxide hydrolase 2;Epoxide hydrolase 2 cytoplasmic;epoxide hydrolase 2, cytosolic;Epoxide hydrolase soluble;HYES_HUMAN;SEH;Soluble epoxide hydrolase;
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
2053
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
OTI1A6
Unconjugated
EPHX2
Liquid
EPHX2
Full length human recombit protein of EPHX2 produced in HEK293T cell
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit