Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tians Epic™ MP Coated Isolation Gowns

Provides protection from nonhazardous liquid splash

$50.95 - $179.00

Specifications

Certifications/Compliance ANSI-ISEA 101-1996
Cuff Style Elastic
Color White
For Use With (Application) Protects against petroleum oils, pesticides, bleach and Sodium Hydroxide
Material Polypropylene
View More Specs

Products 4
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
19-130-3674
View Documents
Tians
816851TSXL
X-Large
Case of 30 for $50.95

Save 24%

Reg : $67.20

The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Learn More
Add to cart
 
19-130-3675
View Documents
Tians
816851TS2XL
2X-Large
Case of 30 for $179.00
Add to cart
 
19-130-3676
View Documents
Tians
816851TS3XL
3X-Large
Case of 30 for $95.40
Add to cart
 
19-130-3678
View Documents
Tians
816851TSL
Large This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

  • MP (Micro Porous) coated isolation gowns are both breathable and comfortable
  • Best used in wet environments where keeping cool is a concern
  • Protects against petroleum oils, pesticides, bleach and sodium hydroxide
  • T-Strap and elastic wrists

Specifications

Specifications

ANSI-ISEA 101-1996
White
Polypropylene
Elastic
Protects against petroleum oils, pesticides, bleach and Sodium Hydroxide
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit