Learn More
(±)-Epichlorohydrin, 99%, Thermo Scientific Chemicals
Description
Epichlorohydrin is used in the manufacture of polyamines and polyquaternary ammonium salts, as flocculents in water and wastewater treatment. It is used in the synthesis of surface active agents for washing products and toiletries. Polyamine-epichlorohydrin resin is used in paper industry to improve paper wet-strength. It acts as a chemical intermediate and widely used in the manufacture of epoxy resins, epichlorohydrin elastomers and polyamide-epichlorohydrin resins.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Epichlorohydrin is used in the manufacture of polyamines and polyquaternary ammonium salts, as flocculents in water and wastewater treatment. It is used in the synthesis of surface active agents for washing products and toiletries. Polyamine-epichlorohydrin resin is used in paper industry to improve paper wet-strength. It acts as a chemical intermediate and widely used in the manufacture of epoxy resins, epichlorohydrin elastomers and polyamide-epichlorohydrin resins.
Solubility
Miscible with water and organic polar solvents.
Notes
Keep away from sources of ignition. Keep container tightly closed in a dry and well-ventilated place.
Specifications
Melting Point | -57°C |
Density | 1.182 |
Boiling Point | 115°C to 117°C |
Flash Point | 32°C (90°F) |
Odor | Like Chlorine |
Assay Percent Range | 99% |
Refractive Index | 1.438 |
Quantity | 100 g |
UN Number | UN2023 |
Beilstein | 79785 |
Show More |
Safety and Handling
Hazard Category | H226-H301+H311+H331-H314-H317-H335-H350 |
Hazard Statement | GHS H Statement H301-H311-H331-H350-H314-H226-H317 Toxic if swallowed. Toxic in contact with skin. Toxic if inhaled. May cause cancer. Causes severe skin burns and eye damage. Flammable liquid and vapour. May cause an allergic skin reaction. |
Precautionary Statement | P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P272-P280g-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P370+P378q-P501c |
DOTInformation | Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: EPICHLOROHYDRIN |
EINECSNumber | 203-439-8 |
RTECSNumber | TX4900000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.