Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Lab Vision™ Epidermal Growth Factor Receptor/EGFR Ab-11, Mouse Monoclonal Antibody

Obtain accurate, reproducible results in western blotting and immunoprecipitation experiments.

$209.00 - $1043.00

Specifications

Antigen Epidermal Growth Factor Receptor/EGFR Ab-11
Clone 199.12
Classification Monoclonal
Conjugate Unconjugated
Format Concentrated
View More Specs

Products 5
Catalog Number Mfr. No. Concentration Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Concentration Quantity Price Quantity & Availability  
MS396P
View Documents
Thermo Scientific™
MS396P
200μg/mL 1mL
Each for $757.50
Add to cart
 
MS396P0
View Documents
Thermo Scientific™
MS396P0
200μg/mL 100μL
Each for $209.00
Add to cart
 
MS396P1
View Documents
Thermo Scientific™
MS396P1
200μg/mL 500μL
Each for $560.50
Add to cart
 
MS396P1ABX
View Documents
Thermo Scientific™
MS396P1ABX
1mg/mL 100μL
Each for $617.50
Add to cart
 
MS396PABX
View Documents
Thermo Scientific™
MS396PABX
1mg/mL 200μL
Each for $1,043.00
Add to cart
 
Description

Description

Obtain accurate, reproducible results in western blotting and immunoprecipitation experiments with Thermo Scientific™ Epidermal Growth Factor Receptor/EGFR Ab-11, Mouse Monoclonal Antibody.

EGFR is a type I receptor tyrosine kinase with sequence homology to erbB-1, -2, -3 -4 or HER-1, -2, -3 -4. It binds to Epidermal Growth Factor (EGF), Transforming Growth Factor-a (TGF-a), Heparin-binding EGF (HB-EGF), amphiregulin, βcellulin and epiregulin. EGFR is overexpressed in tumors of breast, brain, bladder, lung, gastric, head & neck, esophagus, cervix, vulva, ovary, and endometrium. It is predominantly present in squamous cell carcinomas.

Host Species: Mouse

Clone: 199.12

Isotype: IgG2a

Species Reactivity: Human. Others not known.

Epitope: Extracellular domain

Immunogen: Extracellular domain of human recombinant EGFR protein

Molecular Weight: 170 kDa (wild type) and 145 kDa (vIII variant)

Positive Control: A431 cells

Cellular Localization: Membrane

Recommended for:

  • Western Blotting
  • Immunoprecipitation (Native only)
  • Effect on EGF Binding (Noninhibitory)

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

Epidermal Growth Factor Receptor/EGFR Ab-11
Monoclonal
Concentrated
RUO
Primary
IgG2a
199.12
Unconjugated
Murine
EGFR Family
Extracellular domain of human recombinant EGFR protein
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit