Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(-)-Epigallocatechin gallate, Thermo Scientific Chemicals

Catalog No. aaj61745mf
Click to view available options
Quantity:
50 mg

 

Epigallocatechin gallate is reported to strongly inhibit Cu(2+)-mediated oxidative modification of LDLs. EGCG also inhibits NOS2 (iNOS).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Epigallocatechin gallate is reported to strongly inhibit Cu(2+)-mediated oxidative modification of LDLs. EGCG also inhibits NOS2 (iNOS).

Solubility
Soluble in ethanol, dimethyl formamide, water.

Notes
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 989-51-5
Molecular Formula C22H18O11
Molecular Weight (g/mol) 458.375
MDL Number MFCD00075940
InChI Key WMBWREPUVVBILR-WIYYLYMNSA-N
Synonym =--epigallocatechin gallate, egcg, epigallocatechin gallate, epigallocatechin 3-gallate, tea catechin, epigallocatechin-3-gallate, =--epigallocatechin-3-o-gallate, teavigo, =--epigallocatechol gallate, epigallocatechin-3-monogallate
PubChem CID 65064
ChEBI CHEBI:4806
IUPAC Name [(2R,3R)-5,7-dihydroxy-2-(3,4,5-trihydroxyphenyl)-3,4-dihydro-2H-chromen-3-yl] 3,4,5-trihydroxybenzoate
SMILES C1C(C(OC2=CC(=CC(=C21)O)O)C3=CC(=C(C(=C3)O)O)O)OC(=O)C4=CC(=C(C(=C4)O)O)O

Specifications

Melting Point 216°C to 218°C
Color Orange to Pink
Quantity 50 mg
Merck Index 143526
Solubility Information Soluble in ethanol,dimethyl formamide,water.
Formula Weight 458.4
Physical Form Powder
Chemical Name or Material (-)-Epigallocatechin gallate
Hazard Category H312
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P280-P302+P352-P312-P363-P501c
RTECSNumber KB5200000
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.