Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epinephrine Bitartrate, USP, 97-102%, Spectrum™

Click to view available options
Quantity:
5 g
25 g
100 g
1 kg
Packaging:
Amber Glass Bottle
51-42-3
C13H19NO9
333.29
YLXIPWWIOISBDD-NDAAPVSOSA-N
(2R,3R)-2,3-dihydroxybutanedioic acid; 4-[(1R)-1-hydroxy-2-(methylamino)ethyl]benzene-1,2-diol
O[C@H]([C@@H](O)C(O)=O)C(O)=O.CNC[C@H](O)C1=CC=C(O)C(O)=C1
51-42-3
C13H19NO9
333.29
YLXIPWWIOISBDD-NDAAPVSOSA-N
(2R,3R)-2,3-dihydroxybutanedioic acid; 4-[(1R)-1-hydroxy-2-(methylamino)ethyl]benzene-1,2-diol
O[C@H]([C@@H](O)C(O)=O)C(O)=O.CNC[C@H](O)C1=CC=C(O)C(O)=C1

EP135, 51-42-3, C9H13NO3•C4H6O6

Spectrum™ Epinephrine Bitartrate, USP is used to treat life-threatening allergic reactions including anaphylaxis. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Packaging Amber Glass Bottle
Melting Range 147-152°C
Loss on Drying 0.005
Quantity 25 g
Ignition Residue Negligible
CAS Max % 1
UN Number UN2811
Percent Purity 97 to 102%