Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epinephrine, USP, 97-100.5%, Spectrum™

Click to view available options
Quantity:
1 g
5 g
25 g
100 g
1 kg
Packaging:
Amber Glass Bottle
51-43-4
C9H13NO3
183.21
UCTWMZQNUQWSLP-VIFPVBQESA-N
4-[(1R)-1-hydroxy-2-(methylamino)ethyl]benzene-1,2-diol
CNC[C@H](O)C1=CC=C(O)C(O)=C1
51-43-4
C9H13NO3
183.21
UCTWMZQNUQWSLP-VIFPVBQESA-N
4-[(1R)-1-hydroxy-2-(methylamino)ethyl]benzene-1,2-diol
CNC[C@H](O)C1=CC=C(O)C(O)=C1

EP130, 51-43-4, C9H13NO3

Spectrum™ Epinephrine, USP is also known as adrenaline and is a hormone and neurotransmitter. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Packaging Amber Glass Bottle
Loss on Drying 0.02
Quantity 25 g
Ignition Residue Negligible
Specific Rotation -50.0° to -54.0°
CAS Max % 1
UN Number UN2811
Percent Purity 97 to 100.5%