Learn More
Epiregulin Mouse anti-Human, Alexa Fluor™ 350, Clone: MM0265- 12Y4, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP212285AF350
Catalog No. NP21228A30
Add to cart
Description
Description
Epiregulin Monoclonal antibody specifically detects Epiregulin in Human samples. It is validated for Western Blot,Immunohistochemistry,Immunohistochemistry (Paraffin).
Specifications
Specifications
Epiregulin
MM0265- 12Y4
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
epiregulin, ER, proepiregulin
Mouse
IgG1
Store at 4°C in the dark.
Monoclonal
Human
Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Alexa Fluor™ 350
50 mM sodium borate with 0.05% sodium azide
EREG
Human EREG
0.1 mL
Primary
2069