Learn More
Epiregulin Mouse anti-Human, Alexa Fluor 647, Clone: MM0265- 12Y4, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP212285AF647
Catalog No. NB212285647
Add to cart
Description
Description
Epiregulin Monoclonal specifically detects Epiregulin in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
Specifications
Epiregulin
MM0265- 12Y4
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Purified
EREG
Human EREG
Protein G purified
Angiogenesis, Biologically Active Proteins
Primary
2069
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Alexa Fluor 647
50mM Sodium Borate with 0.05% Sodium Azide
epiregulin, ER, proepiregulin
Mouse
IgG1
0.1 ml
Store at 4C in the dark.
Monoclonal
Human