Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epiregulin, Mouse anti-Human, Clone: 870110, R&D Systems™

Mouse Monoclonal Antibody

$119.60 - $304.50

Specifications

Antigen Epiregulin
Clone 870110
Applications Neutralization
Host Species Mouse
Target Species Human
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MAB14253100
View Documents
R&D Systems
MAB14253100
100 ug
Each for $304.50
Add to cart
 
MAB14253SP
View Documents
R&D Systems
MAB14253SP
25 ug
Each for $119.60
Add to cart
 
Description

Description

Epiregulin Monoclonal antibody specifically detects Epiregulin in Human Samples. It is validated for Neutralization.
Specifications

Specifications

Epiregulin
Neutralization
Human
Primary
870110
Mouse
Monoclonal
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit