Learn More
Epiregulin Rat anti-Mouse, Alexa Fluor™ 750, Clone: 6H12, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP221992AF750
Catalog No. NP221992A70
Add to cart
Description
Description
Epiregulin Monoclonal antibody specifically detects Epiregulin in Mouse samples. It is validated for ELISA.
Specifications
Specifications
Epiregulin
6H12
ELISA
epiregulin, ER, proepiregulin
Rat
IgG2
Store at 4°C in the dark.
Monoclonal
Mouse
ELISA
Alexa Fluor™ 750
50 mM sodium borate with 0.05% sodium azide
EREG
This antibody was produced from a hybridoma (mouse myeloma fused with spleen cells from a rat immunized with mouse recombinant protein Epiregulin. Rat monoclonal anti mouse Epiregulin Q61521.
0.1 mL
Primary
2069