Learn More
Epithelial Membrane Antigen/EMA Ab-3 Mouse Monoclonal Antibody, Epredia™
Ensure accurate, reproducible results in immunohistochemistry experiments with Epredia™ Epithelial Membrane Antigen/EMA Ab-3, Mouse Monoclonal Antibody.
$174.00 - $540.00
Specifications
Antigen | Epithelial Membrane Antigen/EMA Ab-3 |
---|---|
Clone | E29 |
Applications | Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Form | Concentration | Quantity | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Form | Concentration | Quantity | Price | Quantity & Availability | |||
MS741P0
|
Epredia
MS741P0 |
Concentrated | 200μg/mL | 100 μL |
Each for $174.00
|
|
|||
MS741P1
|
Epredia
MS741P1 |
Concentrated | 200μg/mL | 500 μL |
Each for $378.00
|
|
|||
MS741P
|
Epredia
MS741P |
Concentrated | 200μg/mL | 1 mL |
Each for $540.00
|
|
|||
MS741P1ABX
|
Epredia
MS741P1ABX |
Concentrated | 1mg/mL | 100 μL |
Each for $393.00
|
|
|||
MS741R7
|
Epredia
MS741R7 |
Prediluted | — | 7 mL |
Each for $384.00
|
|
|||
MS741PABX
|
Epredia
MS741PABX |
Concentrated | 1mg/mL | 200 μL | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||
MS741R1
|
Epredia
MS741R1 |
Prediluted | — | 1 mL |
Item Discontinued
This item has been discontinued by the supplier. Please Sign In to view product availability in your area. View alternate product |
N/A | |||
Description
Epithelial membrane antigen (EMA), is a mucin-like glycoprotein. Antibody to EMA has been shown useful as a pan-epithelial marker for detecting early metastatic loci of carcinoma in the bone marrow or liver.Host Species: Mouse
Clone: E29
Isotype: IgG2a/κ
Species Reactivity: Human. Does not react with rat. Others not known.
Epitope: RPAP
Immunogen: Delipidated extract of human milk fat globule membranes
Molecular Weight: 265-400 kDa
Positive Control: Breast or Colon CA
Cellular Localization: Cytoplasmic and Membrane
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifications
Epithelial Membrane Antigen/EMA Ab-3 | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
IVD | |
Human | |
IgG2a κ |
E29 | |
Monoclonal | |
Mouse | |
Oncogenesis | |
Delipidated extract of human milk fat globule membranes | |
Primary |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.