Learn More
epithelial Sodium Channel beta Mouse anti-Mouse, Alexa Fluor 647, Clone: 7B8, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP259380AF647
Catalog No. NB259380647
Add to cart
Description
Description
epithelial Sodium Channel beta Monoclonal specifically detects epithelial Sodium Channel beta in Mouse samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
Specifications
epithelial Sodium Channel beta
7B8
Western Blot, Immunohistochemistry
Purified
SCNN1B
Synthetic peptide from the C-terminal of Rat ENaC beta (aa. 617-638)
Protein G purified
Phospho Specific
Primary
6338
Mouse
Western Blot, Immunohistochemistry
Alexa Fluor 647
50mM Sodium Borate with 0.05% Sodium Azide
amiloride-sensitive sodium channel subunit beta, amiloride-sensitive sodium channel subunit beta 1, BESC1, Beta-ENaC, beta-NaCH, ENaCB, ENaCbeta, Epithelial Na(+) channel subunit beta, epithelial sodium channel beta-2 subunit, epithelial sodium channel beta-3 subunit, nasal epithelial sodium channel beta subunit, Nonvoltage-gated sodium channel 1 subunit beta, SCNEB, sodium channel, nonvoltage-gated 1, beta
Mouse
IgG1
100 ul
Store at 4C in the dark.
Monoclonal
Detects 87kDa.