Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
epithelial Sodium Channel beta Mouse anti-Mouse, Rat, Biotin, Clone: 140000000000, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP259383B
Description
epithelial Sodium Channel beta Monoclonal specifically detects epithelial Sodium Channel beta in Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
epithelial Sodium Channel beta | |
Monoclonal | |
Biotin | |
PBS with 0.05% Sodium Azide | |
SCNN1B | |
Synthetic peptide from the N-terminal of Rat ENaC alpha (aa. 46-68) | |
100 μL | |
Primary | |
Detects 85kDa. | |
Store at 4C in the dark. | |
IgG1 |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
140000000000 | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
amiloride-sensitive sodium channel subunit beta, amiloride-sensitive sodium channel subunit beta 1, BESC1, Beta-ENaC, beta-NaCH, ENaCB, ENaCbeta, Epithelial Na(+) channel subunit beta, epithelial sodium channel beta-2 subunit, epithelial sodium channel beta-3 subunit, nasal epithelial sodium channel beta subunit, Nonvoltage-gated sodium channel 1 subunit beta, SCNEB, sodium channel, nonvoltage-gated 1, beta | |
Mouse | |
Protein G purified | |
Phospho Specific | |
6338 | |
Mouse, Rat | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title