Learn More
epithelial Sodium Channel beta Mouse anti-Mouse, Rat, Biotin, Clone: 2G4, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP259382B
Catalog No. NBP259382B
Add to cart
Description
Description
epithelial Sodium Channel alpha Monoclonal antibody specifically detects epithelial Sodium Channel alpha in Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
Specifications
epithelial Sodium Channel beta
2G4
Western Blot, Immunohistochemistry
Purified
SCNN1B
Synthetic peptide from the N-terminal of Rat ENaC alpha (aa. 46-68)
Protein G purified
Phospho Specific
Primary
6338
Mouse, Rat
Western Blot, Immunohistochemistry
Biotin
PBS with 0.05% Sodium Azide
amiloride-sensitive sodium channel subunit beta, amiloride-sensitive sodium channel subunit beta 1, BESC1, Beta-ENaC, beta-NaCH, ENaCB, ENaCbeta, Epithelial Na(+) channel subunit beta, epithelial sodium channel beta-2 subunit, epithelial sodium channel beta-3 subunit, nasal epithelial sodium channel beta subunit, Nonvoltage-gated sodium channel 1 subunit beta, SCNEB, sodium channel, nonvoltage-gated 1, beta
Mouse
IgG2b
100 ul
Store at 4C in the dark.
Monoclonal
Detects 85kDa.