Learn More
epithelial Sodium Channel beta Mouse anti-Mouse, Rat, DyLight 405, Clone: 140000000000, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP259383V
Catalog No. NBP259383V
Add to cart
Description
Description
epithelial Sodium Channel beta Monoclonal antibody specifically detects epithelial Sodium Channel beta in Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
epithelial Sodium Channel beta
140000000000
Western Blot, Immunocytochemistry/Immunofluorescence
Purified
SCNN1B
Synthetic peptide from the N-terminal of Rat ENaC alpha (aa. 46-68)
Protein G purified
Phospho Specific
Primary
6338
Mouse, Rat
Western Blot, Immunocytochemistry, Immunofluorescence
DyLight 405
50mM Sodium Borate with 0.05% Sodium Azide
amiloride-sensitive sodium channel subunit beta, amiloride-sensitive sodium channel subunit beta 1, BESC1, Beta-ENaC, beta-NaCH, ENaCB, ENaCbeta, Epithelial Na(+) channel subunit beta, epithelial sodium channel beta-2 subunit, epithelial sodium channel beta-3 subunit, nasal epithelial sodium channel beta subunit, Nonvoltage-gated sodium channel 1 subunit beta, SCNEB, sodium channel, nonvoltage-gated 1, beta
Mouse
IgG1
100 ul
Store at 4C in the dark.
Monoclonal
Detects 85kDa.