Learn More
epithelial Sodium Channel beta Sheep anti-Mouse, Rat, Polyclonal, Novus Biologicals™
Sheep Polyclonal Antibody
Manufacturer:  Novus Biologicals NBP146290
 View more versions of this product
Catalog No. NBP146290
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
epithelial Sodium Channel beta p Ser633 Polyclonal antibody specifically detects epithelial Sodium Channel beta p Ser633 in Mouse, Rat samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
Specifications
epithelial Sodium Channel beta (p Ser633)
LYOPH
Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500
Antisera
SCNN1B
A synthetic peptide to phospho S633 of rat amiloride-sensitive sodium channel subunit beta (SCNN1B, SCNEB) conjugated to an immunogenic carrier protein was used as the immunogen. The antigen is homologous with the corresponding sequence in mouse.
Unpurified
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Polyclonal
Specific for SCNN1B.
Mouse, Rat
Immunohistochemistry, Immunohistochemistry (Paraffin)
Unconjugated
Whole antisera with No Preservative
amiloride-sensitive sodium channel subunit beta, amiloride-sensitive sodium channel subunit beta 1, BESC1, Beta-ENaC, beta-NaCH, ENaCB, ENaCbeta, Epithelial Na(+) channel subunit beta, epithelial sodium channel beta-2 subunit, epithelial sodium channel beta-3 subunit, nasal epithelial sodium channel beta subunit, Nonvoltage-gated sodium channel 1 subunit beta, SCNEB, sodium channel, nonvoltage-gated 1, beta
Sheep
IgG
0.1 ml
Phospho Specific
Primary
6338
Reconstitute with 0.1 ml sterilized water to desired concentration.